Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLNK, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131493

Catalog No. PIPA131493

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Human B-cell Linker Protein (BLNK) exists in two alternatively spliced forms of 68 and 70 kD. It is believed to be the central link between B cell antigen receptor and the downstream effector proteins PLC, Grb2, Vav and Nck. Following activation of B cell

PA1-31493 detects BLNK from human samples.PA1-31493 has been successfully used in Western blot applications.The PA1-31493 immunogen is: Peptide with sequence KDSTRLKYAVKVS, from C Terminus of the protein sequence according to NP_037446
Specifications

Specifications

BLNK
Polyclonal
Unconjugated
BLNK
Purified
BLNK
Peptide with sequence KDSTRLKYAVKVS, from C Terminus of the protein sequence according to NP_037446
IgG fraction
RUO
Antibody
Polyclonal
Human, Murine
Western Blot
0.5mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q8WV28, Q9QUN3
BASH, BLNK-S, LY57, MGC111051, SLP-65, SLP65
Goat
IgG
50μg
Avoid Freeze/Thaw Cycles
Primary
29760
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit