Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blooms Syndrome, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA126718

Catalog No. PIPA126718

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The Bloom syndrome gene product is related to the RecQ subset of DExH box-containing DNA helicases and has both DNA-stimulated ATPase and ATP-dependent DNA helicase activities. Mutations causing Bloom syndrome delete or alter helicase motifs and may disable the 3'-5' helicase activity. The normal protein may act to suppress inappropriate recombination.

PA1-26718 detects Blooms Syndrome Protein Blm from human samples.PA1-26718 has been successfully used in Western blot procedures.The PA1-26718 immunogen is a synthetic peptide corresponding to a portion of human Bloom Syndrome encoded within exon 2.
Specifications

Specifications

Bloom's Syndrome Protein
Polyclonal
Unconjugated
BLM
Purified
BLM
Synthetic peptide corresponding to a portion of human Bloom Syndrome encoded within exon 2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunoprecipitation, Western Blot
1mg/mL
tris citrate, tris phosphate with 0.1% sodium azide; pH 7-8
P54132
BLM
Goat
IgG
50μL
Store at -20°C
Primary
641
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit