Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BLVRB, Mouse, Clone: 2F4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant BLVRB.
Supplier: Abnova Corporation H00000645M09
Description
The final step in heme metabolism in mammals is catalyzed by the cytosolic biliverdin reductase enzymes A and B (EC 1.3.1.24).[supplied by OMIM
Sequence: VACTSAFLLWDPTKVPPRLQAVTDDHIRMHKVLRESGLKYVAVMPPHIGDQPLTGAYTVTLDGRGPSRVISKHDLGHFMLRCLTTDEYDGHSTYPSHQYQSpecifications
BLVRB | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_000713 | |
BLVRB | |
BLVRB (NP_000704, 107 a.a. ∼ 206 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
2F4 | |
Mouse monoclonal antibody raised against a partial recombinant BLVRB. | |
BLVRB | |
BVRB/FLR/MGC117413/SDR43U1 | |
Mouse | |
Affinity chromatography | |
RUO | |
645 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction