Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLZF1, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527918

Catalog No. PIPA527918

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Required for normal Golgi structure and for protein transport from the endoplasmic reticulum (ER) through the Golgi apparatus to the cell surface.

PA5-27918 targets BLZF1 in WB applications and shows reactivity with Rat samples.The PA5-27918 immunogen is recombinant fragment corresponding to a region within amino acids 103 and 383 of Human BLZF1.
Specifications

Specifications

BLZF1
Polyclonal
Unconjugated
BLZF1
Purified
BLZF1
Recombinant fragment corresponding to a region within amino acids 103 and 383 of Human BLZF1
Antigen affinity chromatography
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
8548
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.44mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9H2G9, Q8R2X8
GOLGIN-45, JEM-1, JEM-1s, JEM1, Golgin-45, JEM-1short protein, cytoplasmic protein, p45 basic leucine-zipper nuclear factor
Rabbit
IgG
100μL
Antibody
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit