Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLZF1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526605

Catalog No. PIPA526605

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Required for normal Golgi structure and for protein transport from the endoplasmic reticulum (ER) through the Golgi apparatus to the cell surface.
Specifications

Specifications

BLZF1
Polyclonal
PBS with 0.09% sodium azide
Q9H2G9
GOLGIN-45, JEM-1, JEM-1s, JEM1, Golgin-45, JEM-1short protein, cytoplasmic protein, p45 basic leucine-zipper nuclear factor
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8548
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
BLZF1
Liquid
BLZF1
KLH conjugated synthetic peptide between 156-184 amino acids from the central region of human BLZF1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit