Learn More
Bmi-1 Mouse anti-Human, Mouse, Rat, Rabbit, Clone: F6, MilliporeSigma™ Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05637
Catalog No. 05-637-MI
Add to cart
Description
Description
Anti-Bmi-1 Antibody, clone F6 is a Mouse Monoclonal Antibody for detection of Bmi-1 also known as Polycomb group RING finger protein 4, RING finger protein 51 and has been validated in immunocytochemistry, immunoprecipitation and western blotting.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Bmi-1
F6
Immunoaffinity Purified immunoglobulin in Protein A Purified immunoglobulin in 0.1M Tris-Glycine, 0.15M NaCl, 0.05% sodium azide, pH 7.4.
Purified
Murine
IgG1
100μg
Primary
Human, Murine, Rabbit, Rat
Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
NM_005180
BMI1, MGC12685, PCGF4, flvi-2/bmi-1, RNF51
Immunized with recombinant Bmi-1 protein corresponding to residues 1-202 of mouse Bmi-1.
Protein G purfied
2-8°C for one year
Monoclonal
Documents
Promotions
Promotions
  • MilliporeSigma
    Buy two qualifying MilliporeSigma antibodies and get a third of equal or lesser value at no additional cost.
    01-31-2021