Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMI-1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512197
Description
This antibody is predicted to react with bovine, chicken and mouse based on sequence homology.
BMI is a component of the Polycomb group (PcG) multiprotein PRC1 complex, a complex required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A ′Lys-119′, rendering chromatin heritably changed in its expressibility. In the PRC1 complex, it is required to stimulate the E3 ubiquitin ligase activity of RNF2/RING2. BMI cooperates with the MYC oncogene to produce B-lymphomas.Specifications
BMI-1 | |
Polyclonal | |
BMI1 | |
Polycomb complex protein BMI-1; Polycomb group RING finger protein 4; RING finger protein 51; BMI1; PCGF4, RNF51 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
648 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
P35226 | |
BMI1 | |
KLH conjugated synthetic peptide between 257-286 amino acids from the C-terminal region of human BMI1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title