Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BMP-7 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 8 publications
$184.44 - $1184.00
Specifications
Antigen | BMP-7 |
---|---|
Clone | 164311 |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 1.5-6.0 ug/mL |
Applications | Western Blot, Immunohistochemistry, Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3541SP
|
R&D Systems
MAB3541SP |
25 μg |
Each for $184.44
|
|
|||||
MAB3541100
|
R&D Systems
MAB3541100 |
100 μg |
Each for $532.44
|
|
|||||
MAB3541500
|
R&D Systems
MAB3541500 |
500 μg |
Each for $1,184.00
|
|
|||||
Description
BMP-7 Monoclonal specifically detects BMP-7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
BMP-7 | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 1.5-6.0 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P18075 | |
655 | |
Chinese hamster ovary cell line CHO-derived recombinant human BMP-7 Ser293-His431 Accession # P18075 | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. |
164311 | |
Western Blot, Immunohistochemistry, Neutralization | |
Unconjugated | |
RUO | |
Supplied as a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BMP7, BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1, OP-1OP1, Osteogenic protein 1 | |
BMP7 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human BMP-7 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human (rh) BMP-6 is observed and no cross-reactivity with rhBMP-2, -3, -4, -5, or -8 is observed. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title