Learn More
anti-BMP-9, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP131151
Catalog No. NBP131151
Add to cart
Description
Description
BMP-9 Polyclonal antibody specifically detects BMP-9 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
BMP-9
0.83mg/mL
Western Blot 1:500-1:3000
Affinity Purified
GDF2
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human BMP9. The exact sequence is proprietary.
41 kDa
0.1 ml
Cytokine Research
Primary
2658
Human
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
BMP9BMP-9Bone morphogenetic protein 9, GDF-2, growth differentiation factor 2, growth/differentiation factor 2
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.