Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

bone morphogenetic protein 3, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human BMP3 protein.

Manufacturer:  Abnova Corporation H00000651B01

Catalog No. 89-004-982


Add to cart

Description

Description

BMP3 belongs to the transforming growth factor-beta (TGFB) superfamily. Bone morphogenic protein, also known as osteogenin, induces bone formation. [provided by RefSeq

Sequence: MAGASRLLFLWLGCFCVSLAQGERPKPPFPELRKAVPGDRTAGGGPDSELQPQDKVSEHMLRLYDRYSTVQAARTPGSLEGGSQPWRPRLLREGNTVRSFRAAAAETLERKGLYIFNLTSLTKSENILSATLYFCIGELGNISLSCPVSGGCSHHAQRKHIQIDLSAWTLKFSRNQSQLLGHLSVDMAKSHRDIMSWLSKDITQFLRKAKENEEFLIGFNITSKGRQLPKRRLPFPEPYILVYANDAAISEPESVVSSLQGHRNFPTGTVPKWDSHIRAALSIERRKKRSTGVLLPLQNNELPGAEYQYKKDEVWEERKPYKTLQAQAPEKSKNKKKQRKGPHRKSQTLQFDEQTLKKARRKQWIEPRNCARRYLKVDFADIGWSEWIISPKSFDAYYCSGACQFPMPKSLKPSNHATIQSIVRAVGVVPGIPEPCCVPEKMSSLSILFFDENKNVVLKVYPNMTVESCACR
Specifications

Specifications

bone morphogenetic protein 3
Unconjugated
No additive
NM_001201
BMP-3A
Mouse
50 uL
Cytokines
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human BMP3 protein.
BMP3
Serum
BMP3
BMP3 (NP_001192, 1 a.a. ~ 472 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
651
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit