Learn More
BMP5, Mouse, Clone: 4A7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant BMP5.
Supplier: Abnova Corporation H00000653M88
Description
This gene encodes a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. These proteins are synthesized as prepropeptides, cleaved, and then processed into dimeric proteins. This protein may act as an important signaling molecule within the trabecular meshwork and optic nerve head, and may play a potential role in glaucoma pathogenesis. This gene is differentially regulated during the formation of various tumors. [provided by RefSeq
Sequence: NQNRNKSSSHQDSSRMSSVGDYNTSEQKQACKKHELYVSFRDLGWQDWIIAPEGYAAFYCDGECSFPLNAHMNATNHAIVQTLVHLMFPDHVPKPCCAPTKLNAISVLYFDDSSNVILKKYRNMVVRSCGCHSpecifications
BMP5 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_021073.1 | |
BMP5 | |
BMP5 (NP_066551.1, 323 a.a. ∼ 454 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
4A7 | |
Mouse monoclonal antibody raised against a partial recombinant BMP5. | |
BMP5 | |
MGC34244 | |
Mouse | |
Affinity chromatography | |
RUO | |
653 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.