Learn More
MilliporeSigma™ anti-BMP7, Polyclonal Available on GSA/VA Contract for Federal Government customers only.
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ ST1632100UL
Catalog No. ST1632100UL
Add to cart
Description
Description
Specifically detects BMP7 Clone: in Human samples, and it is validated for Immunoprecipitation
The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-β (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesisin vivo in an extraskeletal site. Based on its expression early in embryogenesis, the BMP encoded by this gene has a proposed role in early development and possible bone inductive activity.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
BMP7
Unconjugated
Unpurified
Full-length human BMP7 (aa 1-412)
100μL
−20°C
Polyclonal
Human
Immunoprecipitation
Undiluted serum.
Rabbit
IgG
RUO
Primary
Recognizes the ∽45-50kDa BMP7 protein.