anti-BMP8B, Clone: 6D6, Abnova™

Mouse Monoclonal

Manufacturer: Abnova Corporation H00000656M01

 View more versions of this product

Catalog No. 89-014-062

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Antigen bone morphogenetic protein 8b
    Format Affinity Purified
    Isotype IgG2a
    Cross Reactivity Human
    Host Species Mouse
    Immunogen BMP8B (NP_001711, 274 a.a. ∽363 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
    Purification Method Protein A column
    Quantity 50μg
    Regulatory Status RUO
    Research Discipline Cytokines
    Primary or Secondary Primary
    Monoclonal or Polyclonal Monoclonal
    Clone 6D6
    Conjugate Unlabeled

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit