Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMPR-IA/ALK-3 Mouse anti-Human, Clone: 87933, R&D Systems™

Mouse Monoclonal Antibody

$122.90

Specifications

Antigen BMPR-IA/ALK-3
Immunogen Mouse myeloma cell line NS0-derived recombinant human BMPR-IA/ALK-3 Met1-Arg152 Accession # CAA80257
Clone 87933
Applications Western Blot
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB2406SP
View Documents
R&D Systems
MAB2406SP
25 ug
Each for $122.90
Add to cart
 
MAB2406100
View Documents
R&D Systems
MAB2406100
100 ug This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
MAB2406500
View Documents
R&D Systems
MAB2406500
500 ug This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

BMPR-IA/ALK-3 Monoclonal antibody MAB2406 Specifically detects BMPR-IA/ALK-3 in Human samples. It is validated for Western Blotting
Specifications

Specifications

BMPR-IA/ALK-3
87933
Unconjugated
CAA80257
Monoclonal
Primary
Detects human BMPR-IA/ALK-3 in direct ELISAs and Western blots. In Western blots, this antibody shows 100% cross-reactivity with recombinant mouse BMPR-IA and no cross-reactivity with recombinant human (rh) BMPR-IB or rhBMPR-II. In direct ELISAs, this antibody does not cross-react with any of these proteins.
Protein A or G purified from hybridoma culture supernatant
Mouse myeloma cell line NS0-derived recombinant human BMPR-IA/ALK-3 Met1-Arg152 Accession # CAA80257
Western Blot
Purified
Mouse
IgG2b
657
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit