Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMPR-IB/ALK-6 Mouse anti-Human, Mouse, Clone: 88614, R&D Systems™

Mouse Monoclonal Antibody

$114.50 - $839.00

Specifications

Antigen BMPR-IB/ALK-6
Immunogen Mouse myeloma cell line NS0-derived recombinant human BMPR-IB/ALK-6
Clone 88614
Applications Western Blot
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB505SP
View Documents
R&D Systems
MAB505SP
25 ug
Each for $114.50
Add to cart
 
MAB505100
View Documents
R&D Systems
MAB505100
100 ug
Each for $377.00
Add to cart
 
MAB505500
View Documents
R&D Systems
MAB505500
500 ug
Each for $839.00
Add to cart
 
Description

Description

BMPR-IB/ALK-6 Monoclonal antibody MAB505 Specifically detects BMPR-IB/ALK-6 in Human, Mouse samples. It is validated for Western Blotting
Specifications

Specifications

BMPR-IB/ALK-6
88614
Unconjugated
Mouse
IgG2a
658
RUO
Mouse myeloma cell line NS0-derived recombinant human BMPR-IB/ALK-6
Western Blot
Purified
Monoclonal
Primary
Detects human and mouse BMPR-IB/ALK-6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse (rm) BMPR-IB and no cross-reactivity with recombinant human (rh) BMPR-IA, rmBMPR-IA, or rhBMPR-II is observed.
Protein A or G purified from ascites
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit