Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BMPR-II Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 5 publications
$172.84 - $1184.00
Specifications
Antigen | BMPR-II |
---|---|
Clone | 73805 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
BMPR-II Monoclonal specifically detects BMPR-II in Human samples. It is validated for Western Blot.Specifications
BMPR-II | |
Western Blot 1 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
Q13873 | |
659 | |
Mouse myeloma cell line NS0-derived recombinant human BMPR-II Ala26-Ile151 Accession # Q13873 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human BMPR-II in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) BMP R-IA or rhBMPR-IB. |
73805 | |
Western Blot | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BMP type II receptor, BMP type-2 receptor, BMPR2, BMPR-2, BMPR3, BMPRII, BMPR-IIprimary pulmonary hypertension 1, Bone morphogenetic protein receptor type II, bone morphogenetic protein receptor type-2, bone morphogenetic protein receptor, type II (serine/threonine kinase), BRK-3, EC 2.7.11, EC 2.7.11.30, FLJ41585, FLJ76945, PPH1BMR2, T-ALK, type II activin receptor-like kinase, type II receptor for bone morphogenetic protein-4 | |
BMPR2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title