Learn More
BNIP1 Mouse anti-Human, Clone: 1G7, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA518492
Catalog No. PIMA518492
Add to cart
Description
Description
Peptide Sequence: MAAPQDVHVR ICNQEIVKFD LEVKALIQDI RDCSGPLSAL TELNTKVKEK FQQLRHRIQD LEQLAKEQDK ESEKQLLLQE VENHKKQMLS NQASWRKANL TCKIAIDNLE KAELLQGGDL LRQRKTTKES LAQTSSTITE SLMGISRMMA QQVQQSEEAM QSLVTSSRTI LDANEEFKSM SGTIQLGRKL ITKYNRRELT DKLLIFLALA LFLATVLYIV KKRLFPFL
This gene is a member of the BCL2/adenovirus E1B 19 kd-interacting protein (BNIP) family. It interacts with the E1B 19 kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19 kDa-like sequences of BCL2, also an apoptotic protector. Alternative splicing of this gene results in four protein products with identical N- and C-termini.
Specifications
Specifications
BNIP1
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q12981
BNIP1; BCL2/adenovirus E1B 19kDa interacting protein 1; NIP1; SEC20; TRG-8; BCL2/adenovirus E1B 19kD interacting protein 1; BCL2/adenovirus E1B 19kD-interacting protein 1
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
662
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
1G7
Unconjugated
BNIP1
Liquid
BNIP1
BNIP1 (AAH10959, 1 a.a. ∽ 228 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human