Learn More
BNIP3 Rabbit anti-Human, DyLight® 488, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522861
Catalog No. PIPA522861
Add to cart
Description
Description
In Western blot a band is seen in hypoxic samples at ~30 and 60 kDa, representing the BNIP3 protein. There are some non-specific bands present as well. It is recommended that normoxic samples are run next to the hypoxic samples as a negative control.
This gene is a member of the BCL2/adenovirus E1B 19 kd-interacting protein (BNIP) family. It interacts with the E1B 19 kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19 kDa-like sequences of BCL2, also an apoptotic protector. This gene contains a BH3 domain and a transmembrane domain, which have been associated with pro-apoptotic function. The dimeric mitochondrial protein encoded by this gene is known to induce apoptosis, even in the presence of BCL2.
Specifications
Specifications
BNIP3
Polyclonal
DyLight™ 488
BNIP3
Liquid
BNIP3
A synthetic peptide against an N-terminal region (within residues 1-50) of the human BNIP3 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
sodium borate with no preservative
Q12983
NIP3, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3
Rabbit
IgG
100 μL
4° C, store in dark
Primary
664