Learn More
Bombesin Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136040
Catalog No. 01-672-229
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A recommended positive control is gut (peripheral nerves), fetal lung.
This gene encodes a member of the bombesin-like family of gastrin-releasing peptides. Its preproprotein, following cleavage of a signal peptide, is further processed to produce either the 27 aa gastrin-releasing peptide or the 10 aa neuromedin C. These smaller peptides regulate numerous functions of the gastrointestinal and central nervous systems, including release of gastrointestinal hormones, smooth muscle cell contraction, and epithelial cell proliferation. These peptides are also likely to play a role in human cancers of the lung, colon, stomach, pancreas, breast, and prostate. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
Specifications
Bombesin
Polyclonal
whole serum with no preservative
P07492, Q8R1I2, P24393
GRP
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
GRP
Liquid
GRP
Synthetic frog bombesin conjugated to bovine thyroglobulin.
250 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
171101, 225642, 2922