Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bombesin Receptor 3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA120717

Catalog No. PIPA120717


Add to cart

Description

Description

PA1-20717 detects Bombesin Receptor 3 in Human samples. PA1-20717 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA1-20717 immunogen corresponds to Synthetic peptide conjugated to KLH..

Bombesin Receptor 3 expression has been reported in lung (normal and cancer), nasal mucosa, placenta, and uterus. ESTs have been isolated from normal brain and kidney cancer libraries. It has a role in sperm cell division, maturation, or function. This receptor mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.
Specifications

Specifications

Bombesin Receptor 3
Polyclonal
Unconjugated
BRS3
Liquid
BRS3
Synthetic peptide conjugated to KLH.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.01% sodium azide; pH 7.7
P32247
RP1-196E23.3
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
680
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit