Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA130762

Catalog No. PIPA130762


Add to cart

Description

Description

PA1-30762 detects Bonzo from human samples. PA1-30762 has been successfully used in Western blot applications. The PA1-30762 immunogen is: Synthetic peptide, corresponding to amino acids 319-338 of Human Bonzo.

Specifications

Specifications

CXCR6
Polyclonal
Unconjugated
CXCR6
Liquid
CXCR6
Synthetic peptide, corresponding to amino acids 319-338 of Human Bonzo.
IgG fraction
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.02% sodium azide
O00574
CD186, STRL33, TYMSTR, C-X-C chemokine receptor type 6, CDw186, CXC-R6, CXCR-6, G protein-coupled receptor, G-protein coupled receptor STRL33, G-protein coupled receptor bonzo
Rabbit
IgG
100 μg
4° C
Primary
10663
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit