Learn More
Borrelia burgdorferi, HRP, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA127289
Catalog No. PIPA127289
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Borrelia burgdorferi is a spirochete and the cause of Lyme disease, a tick transmitted illness of humans and animals. B. burgdorferi may persist in humans and animals for months or years following initial infection, despite a robust humoral immune response. B. burgdorferi resembles other spirochetes in that it is a highly specialized, motile, two-membrane, spiral shaped bacteria which lives primarily as an extracellular pathogen. B. burgdorferi has an unusual genome compared with other eubacteria which includes a linear chromosome approximately one megabase in size and numerous linear and circular plasmids.
PA1-27289 detects Borrelia burgdorferi, Treponema pallidum, B. hermsii, and B. parkerii from bacteria samples.PA1-27289 has been successfully used in immunohistochemistry (paraffin), ICC, and Western blot procedures.The PA1-27289 immunogen is whole cell preparation from B. burgdorferi.Store at 4ºC short term. For extended storage aliquot and store at -20ºC or below. Avoid freeze-thaw cycles.
Specifications
Specifications
Borrelia burgdorferi
Polyclonal
HRP
Purified
Rabbit
IgG
500μL
Store at -20°C
Primary
Bacteria
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1mg/mL
PBS with 10mg/mL BSA and 0.002% thimerosal
B. burgdorferi, Lyme disease, Borrelia
Whole cell preparation from B. burgdorferi.
IgG fraction
RUO
Antibody
Polyclonal