Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-BORRELIA SP AB FITC  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 53300065
Affinity purified polyclonal antibody to Borrelia species made in Goat and labeled with fluorescein isothiocyanate (FITC). Isolated from a serum pool from goats immunized with heat-killed whole cells of Borrelia burgdorferi.
Catalog No. 50-669-24