Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bovine IgG Isotype Control, Biotin, Novus Biologicals™
SDP

Bovine Antibody

Supplier:  Novus Biologicals NBP196907

Catalog No. NBP196907


Add to Cart

Description

Description

IgG specifically detects IgG in Bovine samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

IgG
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
IGHG1
Multi-step
RUO
3500
Reconstitute with 1.0 ml deionized water (or equivalent).
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
IgG
Biotin
Immunoglobulin G, ImmunoglobulinG
Bovine
1 mg
Isotype Control (Primary)
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Bovine IgG and anti-Bovine Serum.
Bovine
Lyophilized
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only