Learn More
anti-Bovine IgG, HRP, Isotype Control, Novus Biologicals™
Bovine Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196756
Catalog No. NBP196756
Add to cart
Description
Description
IgG antibody specifically detects IgG in Bovine samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
IgG
0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with No Preservative
immunoglobulin heavy constant gamma 1 (G1m marker)
Bovine
Ion exchange chromatography
RUO
Isotype Control
3500
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
HRP
Lyophilized
IGHG1
IgG
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti- Bovine IgG and anti-Bovine Serum.
Bovine