Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-BOVINE IGM (MU) AB FITC  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52300292
Affinity purified polyclonal antibody to Bovine IgM heavy chain (mu) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.
Catalog No. 50-668-78