Learn More
Serum Albumin Mouse anti-Bovine, Clone: 5G10, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen HYB2610202
Catalog No. 01-670-122
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
HYB 261-02-02 detects BSA. HYB 261-02-02 has been successfully used in ELISA procedures. HYB 261-02-02 immunogen is BSA isolated from bovine plasma. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Bovine serum albumin (BSA) is the most abundant protein in bovine plasma. BSA is produced in the liver and the concentration of BSA in plasma is 35-50 mg/mL. The molecular mass is 69 kDa.
Specifications
Specifications
Bovine Serum Albumin
Monoclonal
0.94 mg/mL
PBS with 15mM sodium azide; pH 7.4
P02769
BSA
Mouse
IgG1, lambda
200 μL
4° C, store in dark
Primary
280717
ELISA
5G10
Unconjugated
ALB
Liquid
ALB
BSA isolated from bovine plasma.
Protein A
RUO
Antibody
Monoclonal
Bovine