Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brachyury Rabbit anti-Human, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523405

Catalog No. PIPA523405


Add to cart

Description

Description

Brachyury is a 435 amino acid containing embryonic nuclear transcription factor binding to the palindromic T-site. It binds to the T-box in the N-terminus to regulate the genes required for mesodermal differentiation. It is the earliest indicator of mesoderm formation and differentiation. Brachyury is expressed during gastrula and neurula stages in involuting mesoderm and in the notochord.
Specifications

Specifications

Brachyury
Polyclonal
Unconjugated
TBXT
Liquid
TBXT
Synthetic peptide (363 CVSHNASPPTSSSQYPS 378) of Brachyury protein.
Protein A
RUO
Antibody
Polyclonal
Human, Non-human Primate
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1.0 mg/mL
PBS with 0.05% sodium azide
O15178
RP11-459F1.1, TFT, T brachyury homolog, brachyury protein, protein T
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6862
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit