Learn More
BRAF, Mouse, Clone: 3D2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant BRAF.
Supplier: Abnova Corporation H00000673M02
Description
This gene encodes a protein belonging to the raf/mil family of serine/threonine protein kinases. This protein plays a role in regulating the MAP kinase/ERKs signaling pathway, which affects cell division, differentiation, and secretion. Mutations in this gene are associated with cardiofaciocutaneous syndrome, a disease characterized by heart defects, mental retardation and a distinctive facial appearance. Mutations in this gene have also been associated with various cancers, including non-Hodgkin lymphoma, colorectal cancer, malignant melanoma, thyroid carcinoma, nonsmall cell lung carcinoma, and adenocarcinoma of lung. A pseudogene, which is located on chromosome X, has been identified for this gene. [provided by RefSeq
Sequence: FRPADEDHRNQFGQRDRSSSAPNVHINTIEPVNIDDLIRDQGFRGDGGSTTGLSATPPASLPGSLTNVKALQKSPGPQRERKSSSSSEDRNRMKTLGRRDSpecifications
BRAF | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_004333 | |
BRAF | |
BRAF (NP_004324, 346 a.a. ∼ 445 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, In situ PLA, Western Blot | |
3D2 | |
Mouse monoclonal antibody raised against a partial recombinant BRAF. | |
BRAF | |
B-RAF1/BRAF1/FLJ95109/MGC126806/MGC138284/RAFB1 | |
Mouse | |
Affinity chromatography | |
RUO | |
673 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.