Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B-Raf, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514928

Catalog No. PIPA514928

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

BRAF, a member of the RAF subfamily of Ser/Thr protein kinases, is involved in the transduction of mitogenic signals from the cell membrane to the nucleus. It may play a role in the postsynaptic responses of hippocampal neurons. This cytoplasmic protein is expressed in brain and testis. Defects in BRAF are involved in a wide range of cancers including lung cancer and non-Hodgkin lymphoma (NHL). This protein contains 1 zinc-dependent phorbol-ester and DAG binding domain.

This antibody is predicted to react with mouse based on sequence homology.
Specifications

Specifications

B-Raf
Polyclonal
PBS with 0.09% sodium azide
P15056
B-Raf proto-oncogene serine/threonine-protein kinase, p94, v-Raf murine sarcoma viral oncogene homolog B1
Rabbit
IgG
400μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
673
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
BRAF
Liquid
BRAF
KLH conjugated synthetic peptide between 379-408 amino acids from human BRAF
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit