Learn More
BRAF35 Mouse anti-Human, Mouse, Clone: 4.21, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05641
Catalog No. 05-641-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect BRAF35 with Anti-BRAF35 Antibody, clone 4.21 (Mouse Monoclonal Antibody), that has been shown to work in western blotting.
Specifications
Specifications
BRAF35
4.21
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Murine
IgG
200μg
Primary
Human, Murine
Western Blot
Unconjugated
NM_006339
HMG20B, SMARCE1r, pp8857, PP7706, BRAF35, SOXL, FLJ26127, SMARCE1R, BRAF25, HMGX2
GST fusion protein corresponding to residues 100-317 of human BRAF35
Protein G chromatography
−20°C for 24 months.
Monoclonal