Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRAF35 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14192

Catalog No. PA14192


Add to cart

Description

Description

PA1-4192 detects BRAF35 in human samples. PA1-4192 has been successfully used in Western blot procedures. The PA1-4192 immunogen is a synthetic peptide corresponding to residues K(31) Q E R G E G P R A G E K G S H E E E P(50) of the human structural DNA-binding protein BRAF35. This sequence is 80% conserved in mouse.

Mutations of the human BRCA2 gene confer susceptibility to breast cancer. A structural DNA binding component has been identified and is designated as BRCA2-Associated Factor 35 (BRAF35). There is a close association of the BRAF35/BRCA2 complex with condensed chromatin coincident with histone H3 phosphorylation. Experimental evidence suggests a role for the BRCA2/BRAF35 complex in the modulation of cell cycle progression.
Specifications

Specifications

BRAF35
Polyclonal
Unconjugated
HMG20B
Liquid
HMG20B
Synthetic peptide corresponding to residues K(31) Q E R G E G P R A G E K G S H E E E P(50) of human BRAF35.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9P0W2
SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related, SMARCE1-related protein, HMG domain protein HMG
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
10362
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit