Learn More
Brain Derived Neurotrophic Factor Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma AB5613P
Catalog No. AB5613PMI
Add to cart
Description
Description
Anti-Brain Derived Neurotrophic Factor Antibody, pro is an antibody proBDNF for use in Immunohistochemistry and Western Blotting.
Specifications
Specifications
Brain Derived Neurotrophic Factor, Pro
Unconjugated
Affinity Purified
Rabbit
20μg
Primary
NM_001709.3; NM_170731.3; NM_170732.3; NM_170733.2; NM_170734.2; NM_170735.4
Immunohistochemistry, Western Blot
Affinity purified immunoglobulin. Lyophilized from 10mM PBS. Reconstitute with 20μL of sterile distilled water.
BDNF; Abrineurin; neurotrophin; MGC34632
Synthetic peptide from human proBDNF.
Lyophilized: −20°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Polyclonal
Human, Murine, Rat