Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-BRAP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP142695

Catalog No. NBP142695


Add to cart

Description

Description

BRAP Polyclonal antibody specifically detects BRAP in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications

Specifications

BRAP
1.0 mg/mL
Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative)
Affinity Purified
BRAP
The immunogen for this product maps to a region between residue 510 and 560 of human BRCA1 associated protein using the numbering given in entry NP_006759.3 (GeneID 8315).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
BRAP2RING finger protein 52, BRCA1 associated protein, EC 6.3.2.-, IMPBRCA1-associated protein, Impedes mitogenic signal propagation, Renal carcinoma antigen NY-REN-63, RNF52galectin-2-binding protein
Rabbit
IgG
100 ul
Lipid and Metabolism
Primary
8315
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only