Learn More
BRCA2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA185156
Catalog No. PA185156
Add to cart
Description
Description
Predicted to react with cat, chimpanzee, dog, horse, mouse, rat and rhesus monkey based on sequence homology. Heat-mediated antigen retrieval is recommended for the staining of paraffin sections. Rabbit anti Human BRCA2 antibody recognizes human Breast Cancer Type 2 susceptibility protein (BRCA2), a tumour-suppressor protein reported to be involved in double-strand break repair, homologous recombination and S-phase checkpoint activation.
Breast Cancer Type 2 susceptibility protein (BRCA2) is a tumour-suppressor protein reported to be involved in double-strand break repair, homologous recombination and S-phase checkpoint activation.
Specifications
Specifications
BRCA2
Polyclonal
Unconjugated
BRCA2
Liquid
BRCA2
Synthetic peptide corresponding to residues L(2587) I P S N D G K A G K E E F Y(2601) of human BRCA2 precursor protein, conjugated to Keyhole Limet Haemocyanin.
Protein G
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
5 mg/mL
PBS with 0.09% sodium azide
P51587
RP11-298P3.4, BRCC2, BROVCA2, FACD, FAD, FAD1, FANCB, FANCD, FANCD1, GLM3
Rabbit
IgG
1 mL
4°C or -20°C if preferred
Primary
675