Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRCAA1, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA146146

Catalog No. PIPA146146

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The novel BRCAA1 protein's epitope, SSKKQKRSHK, was mapped to the cytoplasm of MCF-7 cells. Analysis has shown that the antigen epitope exhibited positive expression in 65% of breast cancer specimens. Its expression is associated with characteristics tha

Suggested positive control: MCF-7 whole cell lysate.
Specifications

Specifications

BRCAA1
Polyclonal
Unconjugated
ARID4B
Purified
ARID4B
Residues 1046-1055 of the 136 kDa human, mouse and rat BRCAA1 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Western Blot
1mg/mL
PBS with 0.02% sodium azide
Q4LE39, A2CG63, Q9JKB5
RP11-382D8.2, BCAA, BRCAA1, DKFZp313M2420, MGC163290, RBBP1L1, RBP1L1, SAP180
Rabbit
IgG
100μg
Store at -20°C
Primary
51742
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit