Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRD2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA131368

Catalog No. PIPA131368


Add to cart

Description

Description

PA1-31368 detects BRD2 from human samples. PA1-31368 has been successfully used in Western blot applications, where a band of 88 kDa is predicted to be detected. The PA1-31368 immunogen is: Synthetic peptide representing a portion of the protein between the putative nuclear localization signal and the ET domain (LocusLink ID 6046).

This gene encodes a transcriptional regulator that belongs to the BET (bromodomains and extra terminal domain) family of proteins. This protein associates with transcription complexes and with acetylated chromatin during mitosis, and it selectively binds to the acetylated lysine-12 residue of histone H4 via its two bromodomains. The gene maps to the major histocompatability complex (MHC) class II region on chromosome 6p21.3, but sequence comparison suggests that the protein is not involved in the immune response. This gene has been implicated in juvenile myoclonic epilepsy, a common form of epilepsy that becomes apparent in adolescence. Multiple alternatively spliced variants have been described for this gene, but the full-length nature of some of these variants has not been determined.
Specifications

Specifications

BRD2
Polyclonal
Unconjugated
BRD2
Liquid
BRD2
Synthetic peptide representing a portion of the protein between the putative nuclear localization signal and the ET domain (LocusLink ID 6046).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
tris citrate, tris phosphate with 0.1% sodium azide; pH 7-8
P25440
DADB-17J1.3, D6S113E, DKFZp686N0336, FLJ31942, FSH, FSRG1, KIAA9001, NAT, RING3, RNF3
Rabbit
IgG
100 μg
4° C, do not freeze
Primary
6046
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit