Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRD4 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141403

Catalog No. 01-675-532


Add to cart

Description

Description

Suggested positive control: HeLa lysate.

BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; longer variant has a C-terminus region with a praline and glutamine-rich domains. BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.
Specifications

Specifications

BRD4
Polyclonal
Unconjugated
BRD4
Liquid
BRD4
A portion of amino acids 150-200 of human BRD4.
Protein G
RUO
Antibody
Polyclonal
Human, Mouse, Non-human primate, Rat
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
1 mg/mL
PBS with 0.05% sodium azide
O60885, Q9ESU6
HUNK1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
23476, 362844, 57261
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit