Learn More
BRD8 Mouse anti-Human, Clone: 3G8, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA521186
Catalog No. PIMA521186
Add to cart
Description
Description
Peptide Sequence: RSGDQNWVSV SRAIKPFAEP GRPPDWFSQK HCASQYSELL ETTETPKRKR GEKGEVVETV EDVIVRKLTA ERVEELKKVI KETQERYRRL KRDAEL
The protein encoded by this gene interacts with thyroid hormone receptor in a ligand-dependent manner and enhances thyroid hormone-dependent activation from thyroid response elements. This protein contains a bromodomain and is thought to be a nuclear receptor coactivator. Three alternatively spliced transcript variants that encode distinct isoforms have been identified.
Specifications
Specifications
BRD8
Monoclonal
0.5 mg/mL
PBS with no preservative; pH 7.4
Q9H0E9
SMAP, SMAP2, p120, bromodomain-containing protein 8, skeletal muscle abundant protein 2, thyroid hormone receptor coactivating protein of 120 kDa, trCP120
Mouse
IgG2b, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10902
ELISA, Immunocytochemistry, Immunofluorescence
3G8
Unconjugated
BRD8
Liquid
BRD8
BRD8 (NP_631938, 33 a.a. ∽ 128 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human