Learn More
MilliporeSigma™ anti-BrdU, Milli-Mark™, AlexaFluor 488, Clone: Bu-1,
Manufacturer:  MilliporeSigma™ FCMAB101A4
Catalog No. FCMAB101A4
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
BrdU
Bu-1
Purified mouse monoclonal IgG2a supernatant conjugated to Alexa Fluor™ 488 in PBS with 0.1% sodium azide and 15mg/mL BSA. Possible mycoplasma risk contained within this product. Use appropriate measures to prevent cell l
BrdU
Bromodeoxyuridine
Protein G purfied
2-8°C in undiluted aliquots for up to 6 months.
Monoclonal
Flow Cytometry, Immunocytochemistry
Alexa Fluor 488
Purified
Murine
IgG2a
100 Tests
Primary
Human