Learn More
Anti- Brentuximab (Anti-Idiotype) Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Supplier:  R&D Systems FAB9584T100UG
Catalog No. FAB9584T100
Explore available promotions
Add to Cart
This item is not returnable. View return policy
Description
Description
Brentuximab Monoclonal specifically detects Brentuximab in Multi-Species samples. It is validated for Flow Cytometry.
Specifications
Specifications
Brentuximab
Monoclonal
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Full length Brentuximab
100 μg
Primary
Multi-species
Purified
Flow Cytometry
2262C
Flow Cytometry 0.25-1 ug/10^6 cells
Rabbit
Protein A or G purified from cell culture supernatant
RUO
Detects Brentuximab in Western blots and direct ELISAs. In direct ELISAs, no cross-reactivity with Cetuximab, Ibritumomab, Rituximab, Trastuzumab, Tositumomab, Adalimumab, Gemtuzumab, Solanezumab, Teprotumumab, and Aducanumab is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG
Product Suggestions
Product Suggestions