Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brevican Mouse anti-Human, Clone: 450613, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524170

Catalog No. PIMA524170


Add to cart

Description

Description

This antibody also detects a truncated version of Recombinant human Brevican (aa 23-649). Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.

Specifications

Specifications

Brevican
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q96GW7
BEHAB, CSPG7, brain-enriched hyaluro-binding protein, brevican core protein, chondroitin sulfate proteoglycan 7, chondroitin sulfate proteoglycan BEHAB
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
63827
Immunoprecipitation, Neutralization, Western Blot
450613
Unconjugated
BCAN
Lyophilized
BCAN
Mouse myeloma cell line NS0-derived recombit human Brevican isoform 1 Asp23-Pro911
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit