Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-BRIC, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals 46770002
Description
BRIC Polyclonal antibody specifically detects BRIC in Human samples. It is validated for ELISA.Specifications
BRIC | |
Unconjugated | |
20 mM Potassium Phosphate (pH 7.0), 0.15 M NaCl with No Preservative | |
Affinity Purified | |
ATP8B1 | |
This antibody is specific for the N Terminus Region of the target protein. | |
Immunogen affinity purified | |
RUO | |
Primary | |
5205 | |
Human |
ELISA | |
ELISA 1:100 - 1:2000 | |
O43520 | |
ATPase class I type 8B member 1, ATPase, aminophospholipid transporter, class I, type 8B, member 1, ATPase, class I, type 8B, member 1, ATPICPFIC1, BRIC, EC 3.6.3, EC 3.6.3.1, Familial intrahepatic cholestasis type 1, FIC1phospholipid-transporting ATPase IC, PFICE1-E2 ATPase, probable phospholipid-transporting ATPase IC | |
Rabbit | |
IgG | |
0.05 mg | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
Polyclonal | |
This product is specific for Human ATP8B1. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only