Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRMS1L Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523800

Catalog No. PIPA523800


Add to cart

Description

Description

This antibody is predicted to react with bovine based on sequence homology.

The protein encoded by this gene shows sequence similarity to the human breast carcinoma metastasis suppressor (BRMS1) protein and the mammalian Sds3 (suppressor of defective silencing 3) proteins. This protein is a component of the mSin3a family of histone deacetylase complexes (HDAC).
Specifications

Specifications

BRMS1L
Polyclonal
PBS with 0.09% sodium azide
Q5PSV4, Q3U1T3
BRMS1, BRMS1-homolog protein p40, BRMS1-like protein p40, breast cancer metastasis-suppressor 1-like protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
52592, 84312
Western Blot
Unconjugated
BRMS1L
Liquid
BRMS1L
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human BRMS1L
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit