Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRUCE Mouse, Unlabeled, Clone: 4, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611192

 View more versions of this product

Catalog No. BDB611192


Add to cart

Description

Description

Selective proteolysis is essential for the modulation of key cellular processes such as cell cycle progression. However, unlike other post-translational events, proteolysis is irreversible and therefore must occur in unidirectional cellular pathways. In eukaryotes, proteolysis is mediated primarily by the ubiquitin pathway. This pathway designates proteins for degradation by the proteasome, a multicatalytic protease complex. The ubiquitin pathway is a multistep system that tags proteins for degradation via the attachment of ubiquitin molecules to the
target. This attachment is mediated by the ubiquitin activating/conjugating enzymes E1, E2, E3, and BRUCE. BRUCE (BIR Repeat containing Ubiquitin-Conjugating Enzyme) is a novel enzyme that associates with the Golgi and the vesicular system. It contains a UBC (ubiquitin conjugating enzyme) domain, which is essential for catalysis, and a BIR (baculovirus inhibitor of apoptosis repeat) motif. BIR motifs are also found within inhibitor of apoptosis proteins (IAP) and are critical for anti-apoptotic activity. Therefore, BRUCE may function to both mediate ubiquitin-dependent proteolysis and contribute to anti-apoptotic cellular pathways.

Host Species: Mouse
Clone: 4
Isotype: IgG1
Species Reactivity: Human
Immunogen: Mouse BRUCE aa. 372-571
Formula Weight [Chemical]: 528kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

BRUCE
4
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
BIR Repeat containing Ubiquitin-Conjugating Enzyme
Affinity Purified
Mouse BRUCE aa. 372-571
50μg
Cell Biology
Primary
Canine, Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.