Learn More
Brucella abortus Mouse anti-Bacteria, Clone: BA35, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA121682
Catalog No. MA121682
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-21682 detects Brucella abortus in Bacteria samples. MA1-21682 has been successfully used in Western Blot procedures. MA1-21682 immunogen corresponds to Brucella abortus..
Brucella abortus is the causative agent of Brucellosis.
Specifications
Specifications
Brucella abortus
Monoclonal
7.5 mg/mL
PBS with 0.1% sodium azide; pH 7.4
B. abortus; B. abortus Lipopolysaccharide; Brucella abortus lipopolysaccharide
Brucella abortus.
Protein G
RUO
Antibody
Monoclonal
Western Blot
BA35
Unconjugated
Liquid
Mouse
IgG2a
500 μg
4° C, do not freeze
Primary
Bacteria