Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serum Albumin Rabbit anti-Bovine, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523403

Catalog No. PIPA523403

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Bovine serum albumin/bovine albumin/BSA is a serum albumin protein that has numerous biochemical applications including ELISAs (Enzyme-Linked Immunosorbent Assay), immunoblots, and immunohistochemistry. It is also used as a nutrient in cell and microbial culture. In restriction digests, BSA is used to stabilize some enzymes during digestion of DNA and to prevent adhesion of the enzyme to reaction tubes and other vessels. This protein does not affect other enzymes that do not need it for stabilization. BSA is also commonly used to determine the quantity of other proteins, by comparing an unknown quantity of protein to known amounts of BSA. BSA is used because of its stability, its lack of effect in many biochemical reactions, and its low cost since large quantities of it can be readily purified from bovine blood, a byproduct of the cattle industry.
Specifications

Specifications

Bovine Serum Albumin
Polyclonal
Unconjugated
ALB
Liquid
ALB
BSA
Protein A
RUO
Antibody
Polyclonal
Bovine
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P02769
Bovine serum albumin, bovine albumin
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
280717
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit