Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BST-1 Rabbit anti-Human, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523388

Catalog No. PIPA523388


Add to cart

Description

Description

Bone marrow stromal cell antigen 1 (BST1) is a stromal cell line-derived glycosylphosphatidylinositol-anchored molecule that facilitates pre-Bcell growth. BST1, which is also known as ADP-rybosyl cyclase 2 and DAMP-1, is an enzyme which is expressed by BST1 gene in human. BST1 expression is enhanced in bone marrow stromal cell lines derived from patients with rheumatoid arthritis. The polyclonal Bcell abnormalities in rheumatoid arthritis may be, at least in part, attributed to BST1 overexpression in the stromal cell population.
Specifications

Specifications

BST-1
Polyclonal
Unconjugated
BST1
Liquid
BST1
Synthetic peptide (51 CAEYRALLSPEQRNKN 66) of BST1 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Non-human Primate
Immunohistochemistry (Paraffin)
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q10588
Bone marrow stromal cell antigen 1, ADP-ribosyl cyclase 2, BST-1, NAD+ nucleosidase, bone marrow stromal antigen 1, cADPr hydrolase 2, cyclic ADP-ribose hydrolase 2
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
683
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit