Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BST-2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA131166

Catalog No. PIPA131166


Add to cart

Description

Description

PA1-31166 detects BST2 from human samples. PA1-31166 has been successfully used in ELISA, Western blot and dot blot applications. The PA1-31166 immunogen is a synthetic peptide (Human). Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Bone marrow stromal cells are involved in the growth and development of B-cells. The specific function of the protein encoded by the bone marrow stromal cell antigen 2 is undetermined; however, this protein may play a role in pre-B-cell growth and in rheumatoid arthritis.
Specifications

Specifications

BST-2
Polyclonal
Unconjugated
BST2
Liquid
BST2
Synthetic peptide (Human)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Dot Blot, ELISA, Western Blot
0.5 mg/mL
PBS with 0.001% sodium azide; pH 7.2
Q10589
Bone marrow stromal cell antigen 2, Tetherin, PDCA-1, 2310015I10Rik, Bst-2, C87040, DAMP-1, GREG, DAMP-1 protein homolog, HM1.24 antigen, bone marrow stromal antigen 2
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
684
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit