Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTAF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130911
Description
BTAF1 Polyclonal specifically detects BTAF1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
BTAF1 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
BTAF1 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human TAF172. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
9044 | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
1.0mg/mL | |
Western Blot 1:500-1:3000, Immunoprecipitation 1:100-1:500 | |
ATP-dependent helicase BTAF1, BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa (Mot1homolog, S. cerevisiae), B-TFIID transcription factor-associated 170 kDa subunit, EC 3.6.1, EC 3.6.4.-, MOT1MGC138406, TAF(II)170KIAA0940, TAF-172, TAF172BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170 kD (Mot1homolog, S. cerevisiae), TAFII170TATA-binding protein-associated factor 172, TBP-associated factor 172 | |
Rabbit | |
207 kDa | |
100 μL | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction